Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 604385 MIM: 612167 | |||||||||||||||||||||||||||||
Literature Links: |
SCN11A PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SCN11A - sodium voltage-gated channel alpha subunit 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR48 - WD repeat domain 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303402.1 | Intron | NP_001290331.1 | ||||
NM_001303403.1 | Intron | NP_001290332.1 | ||||
NM_020839.3 | Intron | NP_065890.1 | ||||
XM_005265346.3 | Intron | XP_005265403.1 | ||||
XM_011533971.1 | Intron | XP_011532273.1 | ||||
XM_017006960.1 | Intron | XP_016862449.1 |