Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGTATGTTTTTCCTCACTCCTC[A/G]TGACTAGAATAGTGCAGCAAACGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607319 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFMBT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SFMBT1 - Scm-like with four mbt domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016329.3 | 4605 | UTR 3 | NP_057413.2 | |||
XM_005265221.3 | 4605 | UTR 3 | XP_005265278.1 | |||
XM_006713203.3 | 4605 | UTR 3 | XP_006713266.1 | |||
XM_006713204.3 | 4605 | UTR 3 | XP_006713267.1 | |||
XM_011533824.2 | 4605 | UTR 3 | XP_011532126.1 | |||
XM_011533825.2 | 4605 | UTR 3 | XP_011532127.1 |
TMEM110 - transmembrane protein 110 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM110-MUSTN1 - TMEM110-MUSTN1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |