Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607811 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf52 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C6orf52 - chromosome 6 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145020.1 | 306 | Intron | NP_001138492.1 | |||
XM_006715088.3 | 306 | Intron | XP_006715151.1 | |||
XM_011514572.2 | 306 | Intron | XP_011512874.1 | |||
XM_011514574.2 | 306 | Intron | XP_011512876.1 | |||
XM_011514575.2 | 306 | Intron | XP_011512877.1 |
PAK1IP1 - PAK1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017906.2 | 306 | Intron | NP_060376.2 | |||
XM_005249204.2 | 306 | UTR 5 | XP_005249261.1 | |||
XM_011514720.1 | 306 | Missense Mutation | ACC,AGC | T,S 25 | XP_011513022.1 | |
XM_011514721.1 | 306 | Missense Mutation | ACC,AGC | T,S 25 | XP_011513023.1 |
Set Membership: |
HapMap |