Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGAGGATAGAATTCTTTGGTGG[C/T]TCATGCTTCTTTGAATGCCATTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608384 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSDMC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSDMC - gasdermin C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031415.2 | Intron | NP_113603.1 | ||||
XM_011517158.1 | Intron | XP_011515460.1 | ||||
XM_011517159.1 | Intron | XP_011515461.1 | ||||
XM_011517161.1 | Intron | XP_011515463.1 | ||||
XM_011517162.1 | Intron | XP_011515464.1 | ||||
XM_011517163.2 | Intron | XP_011515465.1 | ||||
XM_011517165.1 | Intron | XP_011515467.1 | ||||
XM_011517166.2 | Intron | XP_011515468.1 | ||||
XM_011517167.1 | Intron | XP_011515469.1 | ||||
XM_011517168.1 | Intron | XP_011515470.1 | ||||
XM_017013644.1 | Intron | XP_016869133.1 | ||||
XM_017013645.1 | Intron | XP_016869134.1 |