Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTGATGAGGAGGAGTCAGGACC[A/G]CCAAGGTATTTTACTTCAAACCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607515 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLAC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLAC8 - placenta specific 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130715.1 | Intron | NP_001124187.1 | ||||
NM_001130716.1 | Intron | NP_001124188.1 | ||||
NM_016619.2 | Intron | NP_057703.1 |