Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606449 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTP4A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTP4A3 - protein tyrosine phosphatase type IVA, member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007079.3 | Intron | NP_009010.2 | ||||
NM_032611.2 | Intron | NP_116000.1 | ||||
XM_005250764.2 | Intron | XP_005250821.1 | ||||
XM_006716500.2 | Intron | XP_006716563.1 | ||||
XM_011516803.1 | Intron | XP_011515105.1 | ||||
XM_011516805.1 | Intron | XP_011515107.1 | ||||
XM_017012998.1 | Intron | XP_016868487.1 | ||||
XM_017012999.1 | Intron | XP_016868488.1 |
Set Membership: |
HapMap |