Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614151 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFWD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RFWD3 - ring finger and WD repeat domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018124.3 | 2781 | UTR 3 | NP_060594.3 | |||
XM_005256021.4 | 2781 | UTR 3 | XP_005256078.1 | |||
XM_005256022.4 | 2781 | UTR 3 | XP_005256079.1 | |||
XM_006721228.3 | 2781 | UTR 3 | XP_006721291.1 | |||
XM_011523191.2 | 2781 | UTR 3 | XP_011521493.1 | |||
XM_017023391.1 | 2781 | UTR 3 | XP_016878880.1 | |||
XM_017023392.1 | 2781 | UTR 3 | XP_016878881.1 |
Set Membership: |
HapMap |