Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGGAGGAAATTTTGTTCACTTA[C/G]TAGCTGTATTCCCAGCCCATCCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603024 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID1A - AT-rich interaction domain 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006015.4 | Intron | NP_006006.3 | ||||
NM_139135.2 | Intron | NP_624361.1 |
LOC101928728 - uncharacterized LOC101928728 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |