Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611130 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHMP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHMP7 - charged multivesicular body protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317899.1 | 2039 | Intron | NP_001304828.1 | |||
NM_152272.4 | 2039 | Intron | NP_689485.1 | |||
XM_006716415.2 | 2039 | Intron | XP_006716478.1 | |||
XM_017013961.1 | 2039 | Intron | XP_016869450.1 | |||
XM_017013962.1 | 2039 | Intron | XP_016869451.1 | |||
XM_017013963.1 | 2039 | Intron | XP_016869452.1 | |||
XM_017013964.1 | 2039 | UTR 5 | XP_016869453.1 | |||
XM_017013965.1 | 2039 | Intron | XP_016869454.1 |
Set Membership: |
HapMap |