Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609263 | ||||||||||||||||||||
Literature Links: |
SEH1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEH1L - SEH1 like nucleoporin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013437.1 | Intron | NP_001013455.1 | ||||
NM_031216.3 | Intron | NP_112493.2 | ||||
XM_005258152.4 | Intron | XP_005258209.1 | ||||
XM_011525742.2 | Intron | XP_011524044.1 | ||||
XM_011525743.2 | Intron | XP_011524045.1 | ||||
XM_011525744.1 | Intron | XP_011524046.1 | ||||
XM_011525745.2 | Intron | XP_011524047.1 | ||||
XM_011525746.2 | Intron | XP_011524048.1 | ||||
XM_017026025.1 | Intron | XP_016881514.1 | ||||
XM_017026026.1 | Intron | XP_016881515.1 | ||||
XM_017026027.1 | Intron | XP_016881516.1 |