Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAGTGTGGAGAGCTGAGTATTC[G/T]GAGGTGTTTCATGCCTCATTTTCTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Frmd6 PubMed Links |
Frmd6 - FERM domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_028127.3 | Intron | NP_082403.2 |