Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTTTCAAGGAAAGATACTGGAA[C/T]GTGTGATCCTGGAGTACTTTGCCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Prph2 PubMed Links |
Prph2 - peripherin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008938.1 | Intron | NP_032964.1 |