Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gm16630 PubMed Links |
Gm16630 - predicted gene, 16630 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Krba1 - KRAB-A domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133922.3 | 755 | Missense Mutation | NP_598683.3 |