Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGGCCTGAAAAGTCCCAGGTAG[C/T]TTAAGGGACAAGGGACACCCTGCCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fmnl1 PubMed Links |
Fmnl1 - formin-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077698.1 | Intron | NP_001071166.1 | ||||
NM_019679.2 | Intron | NP_062653.2 |