Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTTCACCTTTCCAACAGATACAG[A/T]CAGAGGAACTCATAACACTGTCCCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Chodl PubMed Links |
Chodl - chondrolectin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139134.3 | 157 | UTR 5 | NP_624360.2 |