Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pnrc2 PubMed Links |
Pnrc2 - proline-rich nuclear receptor coactivator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026383.3 | 518 | Missense Mutation | NP_080659.1 |
Srsf10 - serine/arginine-rich splicing factor 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |