Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCAAGCCTTAGAAAAGGTCATT[A/G]CTGTCCAGAAGAATGCCACCCCTTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dok2 PubMed Links |
Dok2 - docking protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010071.2 | 790 | Missense Mutation | NP_034201.1 |