Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTAGGTGGATCAGCTGATTTATT[C/T]GGGGGATTTGCTGACTTTGGCTCAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Clint1 PubMed Links |
Clint1 - clathrin interactor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001045520.3 | 1220 | Silent Mutation | NP_001038985.2 |