Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGAAAATAAATGTCATGAGAAG[C/T]TACCACCATCTGTACTTCAGTTGTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lama2 PubMed Links |
Lama2 - laminin, alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008481.2 | Intron | NP_032507.2 |