Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCAAGAGGTTGTTCCTGTGATT[C/T]GACTGTAAACCCTCTTAATTTTTAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Map2k4 PubMed Links |
Map2k4 - mitogen-activated protein kinase kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009157.4 | Intron | NP_033183.1 |