Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTATTTATTAAAGGAGAGAACT[G/T]CAAAGGCTGGTCAATATAATGAAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nkain2 PubMed Links |
Nkain2 - Na+/K+ transporting ATPase interacting 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013411.2 | Intron | NP_001013429.2 | ||||
NM_001025286.2 | Intron | NP_001020457.2 |