Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTCAGGTGGAAAGGAGGGACAT[C/T]CTGAAGGTGGGTTCTCAGTGTTCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ddx59 PubMed Links |
Ddx59 - DEAD (Asp-Glu-Ala-Asp) box polypeptide 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026500.3 | 1625 | Silent Mutation | NP_080776.1 |