Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTGTCTTTCAAATTCTATCTTC[A/G]AACAAAAATTGAGACAGACAGACTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
L1cam PubMed Links |
L1cam - L1 cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008478.3 | Intron | NP_032504.3 |