Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGGCGTGAGTCCATTGCAAACA[C/T]TTCTAGTTGAGTGCTCACAGGAGTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Jazf1 PubMed Links |
Jazf1 - JAZF zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168277.1 | Intron | NP_001161749.1 | ||||
NM_173406.3 | Intron | NP_775582.2 |