Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGACTGAATAGAAATGGGCTTCC[C/G]AAAGCCCCGGATAAGCCTGAAAGAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dock4 PubMed Links |
Dock4 - dedicator of cytokinesis 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172803.2 | 1046 | Silent Mutation | NP_766391.2 |