Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Inpp5k PubMed Links |
Inpp5k - inositol polyphosphate 5-phosphatase K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008916.2 | Intron | NP_032942.1 |
Myo1c - myosin IC | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |