Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTTTCTACTGTATATTCACCAG[C/G]TGAGAGAGATTATTTCAATAGTTTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cdh9 PubMed Links |
Cdh9 - cadherin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009869.1 | Intron | NP_033999.1 |