Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATTTTAGAGCGAGCAGGTTCTT[A/C]AAATACTATTATTCACTCTATTCTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Adamtsl1 PubMed Links |
Adamtsl1 - ADAMTS-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029967.3 | Intron | NP_084243.3 |