Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Baiap2l1 PubMed Links |
Baiap2l1 - BAI1-associated protein 2-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Bri3 - brain protein I3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163709.1 | Intron | NP_001157181.1 | ||||
NM_018772.4 | Intron | NP_061242.3 |
Dmr - Dmrt1 related | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |