Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATTCCATTCGGTTCTTCCCAGT[A/G]TGGGATTCAATCTTCCCTGTCCTTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
3110009E18Rik PubMed Links |
3110009E18Rik - RIKEN cDNA 3110009E18 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172074.1 | Intron | NP_001165545.1 | ||||
NM_028439.2 | Intron | NP_082715.1 |
Dbi - diazepam binding inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037999.2 | Intron | NP_001033088.1 | ||||
NM_007830.4 | Intron | NP_031856.1 |