Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Aqp7 PubMed Links |
Aqp7 - aquaporin 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Nfx1 - nuclear transcription factor, X-box binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023739.3 | 4138 | UTR 3 | NP_076228.2 |