Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCCATGGGAGGACTGGGGATGC[A/G]GGTAGGAGGCTTGGATCCATAGACA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sh3bp4 PubMed Links |
Sh3bp4 - SH3-domain binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133816.2 | Intron | NP_598577.1 |