Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCCAGGGCTTGGGATGAGTGGC[A/G]GTGCTGACTTGTTCACTTGGCAGAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dnmt3a PubMed Links |
Dnmt3a - DNA methyltransferase 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271753.1 | 7362 | UTR 3 | NP_001258682.1 | |||
NM_007872.4 | 7362 | UTR 3 | NP_031898.1 | |||
NM_153743.4 | 7362 | UTR 3 | NP_714965.1 |