Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCTCGTCACAGCCTTCAGGCA[A/G]GGTCCCTGCTGGGCTGCAGCCACAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lama5 PubMed Links |
Lama5 - laminin, alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081171.2 | 1858 | Silent Mutation | NP_001074640.1 |