Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGGCAGACACAGCATGTGGCCT[A/G]AATTATCAGCTAGAACTAAAAGCTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Park2 PubMed Links |
Park2 - Parkinson disease (autosomal recessive, juvenile) 2, parkin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016694.3 | Intron | NP_057903.1 |