Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTAGCTTTGGTTATCATTTTGTT[A/G]TTTTTGAATACTTGCCTTATTTGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Wasf1 PubMed Links |
Wasf1 - WAS protein family, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031877.3 | 2363 | UTR 3 | NP_114083.1 |