Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCGCGCTTCTCCTCCACGGTCTC[C/G]AGTTCGTGGTGTAATCTGACAGACA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mtcl1 PubMed Links |
Mtcl1 - microtubule crosslinking factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114098.1 | 1700 | Silent Mutation | NP_001107570.1 | |||
NM_172963.4 | 1700 | Silent Mutation | NP_766551.3 |