Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAATGAAAGATTCTAAGCTCTGG[A/G]GCTTGGGGAAGACGCCTGAGATATA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dlgap1 PubMed Links |
Dlgap1 - discs, large (Drosophila) homolog-associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128180.1 | Intron | NP_001121652.1 | ||||
NM_001128181.1 | Intron | NP_001121653.1 | ||||
NM_027712.3 | Intron | NP_081988.3 | ||||
NM_177639.6 | Intron | NP_808307.2 |