Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTCTGCGGCAAATCCAGGAAA[A/G]CAGTGCAGCCCTCTTTCTACACCGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pde11a PubMed Links |
Pde11a - phosphodiesterase 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081033.1 | Intron | NP_001074502.1 |