Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAAAACCTGCAGTCAGCTGTTAG[T/C]GCAGGAGTAAAATGTTCAGCATCAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mbnl1 PubMed Links |
Mbnl1 - muscleblind-like 1 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253708.1 | Intron | NP_001240637.1 | ||||
NM_001253709.1 | Intron | NP_001240638.1 | ||||
NM_001253710.1 | Intron | NP_001240639.1 | ||||
NM_001253711.1 | Intron | NP_001240640.1 | ||||
NM_001253713.1 | Intron | NP_001240642.1 | ||||
NM_020007.3 | Intron | NP_064391.2 |