Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGTATACACATAATGAACATCT[C/G]CTCATATCTTTGAACAGTATTATTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Uchl5 PubMed Links |
Uchl5 - ubiquitin carboxyl-terminal esterase L5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159866.1 | Intron | NP_001153338.1 | ||||
NM_019562.2 | Intron | NP_062508.2 |