Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTTAAGCACCCTTTCCAATCA[G/T]TCTTATTTAGTGCATTGCCTGTAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
F13b PubMed Links |
F13b - coagulation factor XIII, beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031164.2 | Intron | NP_112441.2 |