Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCTCCAGGCGGGTCTTGTACGT[A/G]AGCAGGTCATCCGTCTGGTTCACGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tekt1 PubMed Links |
Tekt1 - tektin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282006.1 | 383 | Silent Mutation | NP_001268935.1 | |||
NM_001282007.1 | 383 | Silent Mutation | NP_001268936.1 | |||
NM_011569.3 | 383 | Silent Mutation | NP_035699.2 |