Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATAAATATAAGTCAGTACTAAAT[A/G]GGGAGCTAGATACTAACTTCACATT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Crim1 PubMed Links |
Crim1 - cysteine rich transmembrane BMP regulator 1 (chordin like) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015800.3 | Intron | NP_056615.1 |