Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Aco2 PubMed Links |
Aco2 - aconitase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080633.2 | 2279 | Silent Mutation | NP_542364.1 |
Polr3h - polymerase (RNA) III (DNA directed) polypeptide H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030229.4 | 2279 | Intron | NP_084505.2 |