Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGATCAAAGACTACATCATCCC[C/T]GACTTGCTGGGTGGACTCAGTGGAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc26a9 PubMed Links |
Slc26a9 - solute carrier family 26, member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177243.4 | 329 | Silent Mutation | NP_796217.2 |