Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACACTCCTGGCAGCATCCATAAA[C/G]CCAATGGGTATAAACCTCTTTCATG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pdzd2 PubMed Links |
Pdzd2 - PDZ domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081064.1 | Intron | NP_001074533.1 |