Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATGCTGCTGGGGACTACAGGA[A/G]TTGAGCACAGTGGTCTATGAGTTTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc2a12 PubMed Links |
Slc2a12 - solute carrier family 2 (facilitated glucose transporter), member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178934.4 | Intron | NP_849265.2 |