Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATTTCCAGTCCACCATAGGGGA[A/G]AATGTGTCTCCCTGCTGGAATGTGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Agap1 PubMed Links |
Agap1 - ArfGAP with GTPase domain, ankyrin repeat and PH domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037136.1 | Intron | NP_001032213.1 | ||||
NM_178119.3 | Intron | NP_835220.1 |