Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTCTAGACACATTCCCTTCCCC[C/T]AGTCTGAGCATTTCTTTATCTTAAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Stk32a PubMed Links |
Stk32a - serine/threonine kinase 32A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178749.3 | Intron | NP_848864.1 |